This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216636_m1
See other TMPRSS11A GE Assays ›
Gene Symbol:
Tmprss11a
Gene Name:
transmembrane protease, serine 11a
Gene Aliases:
Gm7
Chromosome Location:
Chr.5: 86410410 - 86469175 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216636_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033233.2 NP_001028405.1 3 - 4 341 146
GenBank mRNA AK142820.1 - 3 - 4 342 146

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002681.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Protease Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cell cycle
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500023664.mm.1 NM_001033233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023664.mm.1 NM_001033233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500023664.mm.1 TC0500002681.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023665.mm.1 NM_001033233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023665.mm.1 NM_001033233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500023665.mm.1 TC0500002681.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-t8vjm:80/100.66.78.50:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline