Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 9 - 10 | 950 | 82 | |
| GenBank mRNA | AK144802.1 | - | 9 - 10 | 932 | 82 | |
| AK150237.1 | - | 9 - 10 | 929 | 82 | ||
| AK168215.1 | - | 9 - 10 | 932 | 82 | ||
| AK170722.1 | - | 9 - 10 | 932 | 82 | ||
| AK196574.1 | - | 2 - 3 | 120 | 82 | ||
| AK201678.1 | - | 2 - 3 | 83 | |||
| BC010211.1 | - | 9 - 10 | 928 | 3969515 | 82 | |
| BC024113.1 | - | 5 - 6 | 422 | 5097944 | 82 | |
| BC027006.1 | - | 5 - 6 | 470 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map