This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217225_m1
See other NELFA GE Assays ›
Gene Symbol:
Nelfa
Gene Name:
negative elongation factor complex member A, Whsc2
Gene Aliases:
Whsc2, Whsc2h
Chromosome Location:
Chr.5: 33898179 - 33936258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217225_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nelfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011914.2 NP_036044.1 3 - 4 572 67
GenBank mRNA AK019191.1 - 3 - 4 637 67
AK078760.1 - 3 - 4 572 67
AK088384.1 - 3 - 4 588 67
AK144173.1 - 586 67
AK146185.1 - 3 - 4 624 67
BC038003.1 - 3 - 4 573 4912719 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002223.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nelfa

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020318.mm.1 NM_011914.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020318.mm.1 NM_011914.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500020318.mm.1 TC0500002223.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0