This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217228_m1
See other NELFA GE Assays ›
Gene Symbol:
Nelfa
Gene Name:
negative elongation factor complex member A, Whsc2
Gene Aliases:
Whsc2, Whsc2h
Chromosome Location:
Chr.5: 33898179 - 33936258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217228_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nelfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011914.2 NP_036044.1 7 - 8 950 59
GenBank mRNA AF101435.1 - 7 - 8 908 59
AK019191.1 - 7 - 8 1016 59
AK078760.1 - 7 - 8 950 59
AK088384.1 - 7 - 8 966 59
AK144173.1 - 964 59
AK146185.1 - 7 - 8 1002 59
BC038003.1 - 7 - 8 951 4912719 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002223.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nelfa

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020314.mm.1 NM_011914.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020314.mm.1 NM_011914.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500020314.mm.1 TC0500002223.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline