This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217622_m1
See other UBE2K GE Assays ›
Gene Symbol:
Ube2k
Gene Name:
ubiquitin-conjugating enzyme E2K
Gene Aliases:
AW492011, D5Ertd601e, E2-25k, Hip2, Hypg, Lig
Chromosome Location:
Chr.5: 65537238 - 65598989 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217622_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016786.3 NP_058066.2 3 - 4 439 130
GenBank mRNA AB011081.1 - 3 - 4 377 130
AK018366.1 - 3 - 4 413 130
AK035371.1 - 3 - 4 336 130
AK036068.1 - 3 - 4 380 130
AK048194.1 - 3 - 4 430 130
AK139564.1 - 3 - 4 456 130
AK160728.1 - 3 - 4 399 130
AK162142.1 - 3 - 4 380 130
AK166647.1 - 3 - 4 348 130
BC002013.1 - 3 - 4 287 3496393 130
BC085311.1 - 3 - 4 365 30148009 130

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK035371.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2k

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein ubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
protein K48-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500005474.mm.1 TC0500000614.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005474.mm.1 NM_016786.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0