This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217730_m1
See other BRE GE Assays ›
Gene Symbol:
Bre
Gene Name:
brain and reproductive organ-expressed protein
Gene Aliases:
6030405P19Rik, AI429776, B830038C02Rik
Chromosome Location:
Chr.5: 31697945 - 32084948 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217730_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144541.1 NP_653124.1 5 - 6 550 113
NM_181279.1 NP_851796.1 7 - 8 661 113
NM_181280.1 NP_851797.1 6 - 7 559 113
NM_181281.1 NP_851798.1 4 - 5 394 113
NM_181282.1 NP_851799.1 5 - 6 471 113
GenBank mRNA AF440752.1 - 5 - 6 550 113
AF527952.1 - 7 - 8 661 113
AF527953.1 - 5 - 6 471 113
AF527954.1 - 5 - 6 481 113
AF527955.1 - 4 - 5 394 113
AF538925.1 - 6 - 7 559 113
AK050695.1 - 5 - 6 595 113
AK080991.1 - 5 - 6 586 113
AK156929.1 - 5 - 6 642 113
AK166884.1 - 5 - 6 586 113
BC061000.1 - 5 - 6 626 30305062 113
BC100565.1 - 1 - 2 105 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003709.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bre

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
apoptotic process
cellular response to DNA damage stimulus
response to ionizing radiation
chromatin modification
G2 DNA damage checkpoint
positive regulation of DNA repair
tumor necrosis factor receptor binding
polyubiquitin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_144541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_144541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181280.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181281.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181282.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002996.mm.1 NM_181282.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_144541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_144541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181280.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181281.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181282.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002995.mm.1 NM_181282.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0