This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217842_g1
See other CYTL1 GE Assays ›
Gene Symbol:
Cytl1
Gene Name:
cytokine-like 1
Gene Aliases:
4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147
Chromosome Location:
Chr.5: 37735519 - 37739820 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217842_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cytl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081106.1 NP_001074575.1 2 - 3 229 55
GenBank mRNA AK076770.1 - 1 - 2 121 55
BC063103.1 - 2 - 3 189 55
BC116224.1 - 2 - 3 203 40091368 55
BC116225.1 - 2 - 3 203 40091369 55
EF108311.1 - 2 - 3 200 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000425.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s)

chondrocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
inner ear development
chondroitin sulfate proteoglycan biosynthetic process
positive regulation of sequence-specific DNA binding transcription factor activity
cartilage homeostasis

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500004242.mm.1 NM_001081106.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004242.mm.1 NM_001081106.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004242.mm.1 TC0500000425.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline