This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217919_m1
See other ARHGAP22 GE Assays ›
Gene Symbol:
Arhgap22
Gene Name:
Rho GTPase activating protein 22
Gene Aliases:
B230341L19Rik, RHOGAP2, p68RacGAP
Chromosome Location:
Chr.14: 33214003 - 33369936 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217919_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC038272.1 - 1 - 2 177 4237331 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000358.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400003167.mm.1 TC1400000358.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400003168.mm.1 NM_153800.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003168.mm.1 NM_153800.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400003168.mm.1 TC1400000358.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline