This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218026_m1
See other CACFD1 GE Assays ›
Gene Symbol:
Cacfd1
Gene Name:
calcium channel flower domain containing 1
Gene Aliases:
5930434B04Rik, mFwe
Chromosome Location:
Chr.2: 27009841 - 27021089 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218026_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cacfd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243239.1 NP_001230168.1 1 - 2 181 63
NM_001243240.1 NP_001230169.1 1 - 2 181 63
NM_029862.4 NP_084138.1 1 - 2 181 63
GenBank mRNA AK020041.1 - 1 - 2 165 63
AK034046.1 - 1 - 2 165 63
AK038728.1 - 1 - 2 127 63
AK039867.1 - 1 - 2 158 63
AK040379.1 - 1 - 2 88 63
AK043562.1 - 1 - 2 127 63
AK052562.1 - 1 - 2 142 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000462.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cacfd1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
synaptic vesicle endocytosis
molecular_function
calcium channel activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_001243239.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_001243239.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_001243240.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_001243240.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_029862.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 NM_029862.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200003089.mm.1 TC0200000462.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_001243239.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_001243239.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_001243240.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_001243240.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_029862.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 NM_029862.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200003090.mm.1 TC0200000462.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_001243239.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_001243239.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_001243240.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_001243240.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_029862.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 NM_029862.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200003094.mm.1 TC0200000462.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 541496 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline