This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218180_g1
See other CREB3L4 GE Assays ›
Gene Symbol:
Creb3l4
Gene Name:
cAMP responsive element binding protein 3-like 4
Gene Aliases:
1700012K17Rik, 5330432F22Rik, AIBZIP, AV040530, AV258827, CREB4, JAL, TISP40, acre1
Chromosome Location:
Chr.3: 90237500 - 90243524 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218180_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB052779.1 - 2 - 3 203 61
AB182651.1 - 2 - 3 321 61
AK019928.1 - 2 - 3 321 61
AK088919.1 - 2 - 3 220 61
AY049978.1 - 2 - 3 242 61
BC022605.1 - 2 - 3 177 4164994 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002367.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300018363.mm.1 NM_030080.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300018363.mm.1 TC0300002367.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300018365.mm.1 NM_030080.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300018365.mm.1 TC0300002367.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300018366.mm.1 TC0300002367.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline