This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218181_g1
See other CREB3L4 GE Assays ›
Gene Symbol:
Creb3l4
Gene Name:
cAMP responsive element binding protein 3-like 4
Gene Aliases:
1700012K17Rik, 5330432F22Rik, AIBZIP, AV040530, AV258827, CREB4, JAL, TISP40, acre1
Chromosome Location:
Chr.3: 90237500 - 90243524 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218181_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB047647.1 - 4 - 5 508 88
AB052779.1 - 6 - 7 765 88
AB182651.1 - 6 - 7 883 88
AB182652.1 - 4 - 5 508 88
AF287260.1 - 4 - 5 646 88
AK005918.1 - 4 - 5 665 88
AK019928.1 - 6 - 7 883 88
AK088919.1 - 6 - 7 782 88
AY049978.1 - 6 - 7 804 88
BC022605.1 - 6 - 7 739 4164994 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline