This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218276_m1
See other SSR2 GE Assays ›
Gene Symbol:
Ssr2
Gene Name:
signal sequence receptor, beta
Gene Aliases:
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta
Chromosome Location:
Chr.3: 88579584 - 88588420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218276_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025448.3 NP_079724.1 3 - 4 574 73
GenBank mRNA AK005340.1 - 4 - 5 445 73
AK007538.1 - 1 - 2 41 73
AK007720.1 - 1 - 2 41 73
AK010565.1 - 4 - 5 412 73
AK050505.1 - 3 - 4 575 73
AK084530.1 - 4 - 5 412 73
AK214352.1 - 1 - 2 122 73
BC010214.1 - 3 - 4 558 3497894 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000731.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300005176.mm.1 NM_025448.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300005176.mm.1 NM_025448.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300005176.mm.1 TC0300000731.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0