This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218289_g1
See other PAQR6 GE Assays ›
Gene Symbol:
Paqr6
Gene Name:
progestin and adipoQ receptor family member VI
Gene Aliases:
1500001B10Rik
Chromosome Location:
Chr.3: 88364072 - 88368541 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218289_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Paqr6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198410.3 NP_940802.1 2 - 3 113 71
GenBank mRNA AK139144.1 - 2 - 3 114 71
AY424295.1 - 2 - 3 114 71
BC120512.1 - 1 - 2 76 40129435 71
BC125428.1 - 1 - 2 76 40129943 71
BC144954.1 - 1 - 2 76 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000725.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Paqr6

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300005131.mm.1 NM_198410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300005131.mm.1 NM_198410.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300005131.mm.1 TC0300000725.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0