This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218439_g1
See other UVSSA GE Assays ›
Gene Symbol:
Uvssa
Gene Name:
UV stimulated scaffold protein A
Gene Aliases:
4933407H18Rik, D330017J19Rik, Kiaa1530, mKIAA1530
Chromosome Location:
Chr.5: 33378655 - 33419754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218439_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uvssa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081101.2 NP_001074570.1 11 - 12 2083 70
GenBank mRNA AK016722.1 - 10 - 11 1744 70
AK129384.1 - 10 - 11 1663 70
BC061483.1 - 6 - 7 838 70
BC137803.1 - 11 - 12 2007 8860822 70
BC144920.1 - 11 - 12 1922 9053468 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription-coupled nucleotide-excision repair
cellular response to DNA damage stimulus
response to UV
protein ubiquitination
RNA polymerase II core binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0