This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218442_g1
See other UVSSA GE Assays ›
Gene Symbol:
Uvssa
Gene Name:
UV stimulated scaffold protein A
Gene Aliases:
4933407H18Rik, D330017J19Rik, Kiaa1530, mKIAA1530
Chromosome Location:
Chr.5: 33378655 - 33419754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218442_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uvssa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081101.2 NP_001074570.1 1 - 2 303 77
GenBank mRNA AK040506.1 - 1 - 2 306 77
BB590196.2 - 1 - 2 305 77
BC137803.1 - 1 - 2 227 8860822 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC137803.1, NM_001081101.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription-coupled nucleotide-excision repair
cellular response to DNA damage stimulus
response to UV
protein ubiquitination
RNA polymerase II core binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003312.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003312.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003312.mm.1 NM_001081101.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003315.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003315.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003315.mm.1 NM_001081101.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003310.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003310.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003310.mm.1 NM_001081101.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003311.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003311.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003311.mm.1 NM_001081101.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003313.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003313.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003313.mm.1 NM_001081101.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 402620 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0