This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218443_m1
See other UVSSA GE Assays ›
Gene Symbol:
Uvssa
Gene Name:
UV stimulated scaffold protein A
Gene Aliases:
4933407H18Rik, D330017J19Rik, Kiaa1530, mKIAA1530
Chromosome Location:
Chr.5: 33378655 - 33419754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218443_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uvssa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081101.2 NP_001074570.1 2 - 3 406 104
GenBank mRNA AK016722.1 - 1 - 2 67 104
AK028507.1 - 1 - 2 776 104
AK156117.1 - 1 - 2 565 104
BB590196.2 - 2 - 3 408 104
BC137803.1 - 2 - 3 330 8860822 104
BC144920.1 - 2 - 3 245 9053468 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK016722.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription-coupled nucleotide-excision repair
cellular response to DNA damage stimulus
response to UV
protein ubiquitination
RNA polymerase II core binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003316.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003316.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003316.mm.1 NM_001081101.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003317.mm.1 TC0500000357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003317.mm.1 NM_001081101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003317.mm.1 NM_001081101.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0