This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218521_m1
See other STARD13 GE Assays ›
Gene Symbol:
Stard13
Gene Name:
StAR-related lipid transfer (START) domain containing 13
Gene Aliases:
DLC2, GT650
Chromosome Location:
Chr.5: 151037515 - 151428256 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218521_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163493.1 NP_001156965.1 5 - 6 1843 128
NM_146258.2 NP_666370.3 5 - 6 1843 128
GenBank mRNA AK050259.1 - 5 - 6 1844 128
AK136202.1 - 1 - 2 369 128
BC027830.1 - 5 - 6 1855 5320966 128
DV651942.1 - 1 - 2 102 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003694.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard13

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
signal transduction
GTPase activator activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500033744.mm.1 TC0500003694.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033744.mm.1 NM_001163493.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033744.mm.1 NM_001163493.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033744.mm.1 NM_146258.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033744.mm.1 NM_146258.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500033743.mm.1 TC0500003694.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033743.mm.1 NM_001163493.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033743.mm.1 NM_001163493.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0