This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218948_m1
See other MAPK8 GE Assays ›
Gene Symbol:
Mapk8
Gene Name:
mitogen-activated protein kinase 8
Gene Aliases:
AI849689, JNK, JNK1, Prkm8, SAPK1
Chromosome Location:
Chr.14: 33377898 - 33447214 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218948_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapk8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016700.4 NP_057909.1 8 - 9 1065 114
GenBank mRNA AB005663.1 - 7 - 8 898 114
AB206575.1 - 7 - 8 840 114
AB219977.1 - 7 - 8 874 114
AK043102.1 - 8 - 9 1065 114
AK139498.1 - 8 - 9 1053 114
BC053027.1 - 8 - 9 1035 5694771 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001711.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mapk8

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

ossification
regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
response to osmotic stress
signal transduction
JNK cascade
JUN phosphorylation
response to heat
response to UV
regulation of gene expression
positive regulation of gene expression
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of cell migration
regulation of histone deacetylation
positive regulation of microtubule polymerization
neuron projection development
positive regulation of cyclase activity
negative regulation of protein binding
regulation of protein localization
response to hydrogen peroxide
regulation of circadian rhythm
negative regulation of apoptotic process
positive regulation of DNA replication
response to cadmium ion
determination of dorsal identity
rhythmic process
positive regulation of protein metabolic process
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
cellular response to nitric oxide
positive regulation of deacetylase activity
type B pancreatic cell apoptotic process
neuronal stem cell population maintenance
programmed necrotic cell death
positive regulation of determination of dorsal identity
positive regulation of apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
JUN kinase activity
MAP kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
enzyme binding
histone deacetylase regulator activity
histone deacetylase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013331.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013331.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013331.mm.1 TC1400001711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013329.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013329.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013329.mm.1 TC1400001711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0