This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218955_m1
See other MAPK8 GE Assays ›
Gene Symbol:
Mapk8
Gene Name:
mitogen-activated protein kinase 8
Gene Aliases:
AI849689, JNK, JNK1, Prkm8, SAPK1
Chromosome Location:
Chr.14: 33377898 - 33447214 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218955_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapk8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK139498.1 - 11 - 12 1315 80
BC053027.1 - 11 - 12 1297 5694771 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001711.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013322.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013322.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013322.mm.1 TC1400001711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013320.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013320.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013320.mm.1 TC1400001711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013319.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013319.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013319.mm.1 TC1400001711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013317.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013317.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013317.mm.1 TC1400001711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013318.mm.1 NM_016700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013318.mm.1 NM_016700.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013318.mm.1 TC1400001711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0