This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219059_g1
See other SLC7A1 GE Assays ›
Gene Symbol:
Slc7a1
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
Gene Aliases:
4831426K01Rik, AI447493, Atrc-1, Atrc1, Cat1, Rec-1, Rev-1, mCAT-1
Chromosome Location:
Chr.5: 148327410 - 148399904 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219059_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301424.1 NP_001288353.1 11 - 12 1821 70
NM_007513.4 NP_031539.3 11 - 12 1937 70
GenBank mRNA AK139342.1 - 11 - 12 1913 70
AK147991.1 - 11 - 12 1846 70
BC068234.1 - 11 - 12 1828 70
BC145779.1 - 10 - 11 1742 40131082 70
BC145781.1 - 10 - 11 1742 40131084 70
JN226407.1 - 9 - 10 1657 70
JN226408.1 - 9 - 10 1657 70
JN226410.1 - 9 - 10 1657 70
M26687.1 - 11 - 12 1855 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003637.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a1

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
arginine transport
regulation of TOR signaling
L-arginine import across plasma membrane
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500033377.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033377.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033377.mm.1 NM_007513.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500033376.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033376.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033376.mm.1 NM_007513.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500033375.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033375.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033375.mm.1 NM_007513.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0