This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219235_m1
See other PNISR GE Assays ›
Gene Symbol:
Pnisr
Gene Name:
PNN interacting serine/arginine-rich
Gene Aliases:
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18
Chromosome Location:
Chr.4: 21847583 - 21876475 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219235_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnisr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025669.1 NP_079945.1 2 - 3 655 72
GenBank mRNA AK012036.1 - 2 - 3 198 72
AK017507.1 - 2 - 3 182 72
AK020035.1 - 2 - 3 183 72
AK031964.1 - 2 - 3 191 72
AK077040.1 - 1 - 2 377 72
AK081407.1 - 2 - 3 181 72
AK083021.1 - 2 - 3 181 72
AK160288.1 - 2 - 3 181 72
AK162695.1 - 2 - 3 208 72
BC060111.1 - 2 - 3 165 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000183.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001199.mm.1 NM_025669.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001199.mm.1 NM_025669.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001199.mm.1 TC0400000183.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 568747 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0