This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219236_m1
See other PNISR GE Assays ›
Gene Symbol:
Pnisr
Gene Name:
PNN interacting serine/arginine-rich
Gene Aliases:
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18
Chromosome Location:
Chr.4: 21847583 - 21876475 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219236_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnisr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025669.1 NP_079945.1 3 - 4 842 149
GenBank mRNA AK012036.1 - 3 - 4 385 149
AK017507.1 - 3 - 4 369 149
AK020035.1 - 3 - 4 370 149
AK031964.1 - 3 - 4 378 149
AK077040.1 - 2 - 3 151
AK083021.1 - 3 - 4 368 149
AK160288.1 - 3 - 4 368 149
AK162695.1 - 3 - 4 395 149
BC060111.1 - 3 - 4 352 149
BC141031.1 - 3 - 4 706 9055643 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000183.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001206.mm.1 NM_025669.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001206.mm.1 NM_025669.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001206.mm.1 TC0400000183.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400001205.mm.1 NM_025669.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001205.mm.1 NM_025669.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001205.mm.1 TC0400000183.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0