This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219251_m1
See other CDH17 GE Assays ›
Gene Symbol:
Cdh17
Gene Name:
cadherin 17
Gene Aliases:
HPT-1, HPT-1/LI
Chromosome Location:
Chr.4: 11758157 - 11817905 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219251_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019753.4 NP_062727.1 2 - 3 199 70
GenBank mRNA AF177669.1 - 2 - 3 95 70
AK089791.1 - 2 - 3 165 70
AK155633.1 - 2 - 3 200 70
AK171733.1 - 2 - 3 122 70
BC042891.1 - 2 - 3 117 4208360 70
D87912.1 - 2 - 3 144 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000093.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh17

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

transport
oligopeptide transport
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
oligopeptide transmembrane transport
proton-dependent oligopeptide secondary active transmembrane transporter activity
calcium ion binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400000651.mm.1 NM_019753.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000651.mm.1 NM_019753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400000651.mm.1 TC0400000093.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2tqsm:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0