This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220205_g1
See other ETFDH GE Assays ›
Gene Symbol:
Etfdh
Gene Name:
electron transferring flavoprotein, dehydrogenase
Gene Aliases:
0610010I20Rik, AV001013
Chromosome Location:
Chr.3: 79603788 - 79628767 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220205_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etfdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025794.2 NP_080070.2 2 - 3 386 65
GenBank mRNA AK002483.1 - 2 - 3 339 65
AK028686.1 - 2 - 3 338 65
AK037969.1 - 2 - 3 303 65
AK075673.1 - 2 - 3 337 65
AK085493.1 - 2 - 3 355 65
AK141684.1 - 2 - 3 340 65
AK152624.1 - 2 - 3 355 65
AK169686.1 - 2 - 3 291 65
BC012522.1 - 2 - 3 335 2648103 65
BC057670.1 - 2 - 3 353 2810976 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC057670.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etfdh

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
response to oxidative stress
electron transport chain
fatty acid beta-oxidation using acyl-CoA dehydrogenase
oxidation-reduction process
electron-transferring-flavoprotein dehydrogenase activity
electron carrier activity
oxidoreductase activity
metal ion binding
quinone binding
ubiquinone binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300016512.mm.1 NM_025794.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300016512.mm.1 NM_025794.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300016512.mm.1 TC0300002199.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300016517.mm.1 TC0300002199.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0