This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220302_m1
See other ASIC5 GE Assays ›
Gene Symbol:
Asic5
Gene Name:
acid-sensing (proton-gated) ion channel family member 5
Gene Aliases:
Accn5, Blinac, Inac
Chromosome Location:
Chr.3: 81982293 - 82021233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220302_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021370.2 NP_067345.1 7 - 8 1089 92
GenBank mRNA AK038321.1 - 6 - 7 1141 92
Y19035.1 - 7 - 8 1088 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000636.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic5

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
proton transport
sodium ion transmembrane transport
sodium channel activity
hydrogen ion channel activity
ligand-gated sodium channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300004180.mm.1 NM_021370.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300004180.mm.1 NM_021370.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300004180.mm.1 TC0300000636.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0