This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01221761_m1
See other ADGRF3 GE Assays ›
Gene Symbol:
Adgrf3
Gene Name:
adhesion G protein-coupled receptor F3
Gene Aliases:
Gm1041, Gpr113, Pgr23
Chromosome Location:
Chr.5: 30193431 - 30205754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01221761_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014394.2 NP_001014416.2 13 - 14 2966 76
GenBank mRNA AY701869.1 - 13 - 14 2966 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002155.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrf3

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058947 NM_001014394.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058947 AY701869.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933867 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178014 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440528 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495233 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787995 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913913 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633627 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139905 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112577 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163515 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058947 NM_001014394.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058947 AY701869.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P23674 NM_001014394.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P23674 AY255593.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P23674 AY701869.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571344 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571343 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571337 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571348 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571353 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571360 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571355 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571359 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571338 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571352 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571340 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571346 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571351 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571362 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571356 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23571363 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163515 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112577 - -
Whole Mouse Genome G4122A A_52_P23674 NM_001014394.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P23674 AY255593.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P23674 AY701869.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112577 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571298 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23571365 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571340 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04638033 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571369 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773089 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571310 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04715757 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571353 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04157263 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04579543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571261 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571372 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04873613 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04947437 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773079 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571265 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571305 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04938079 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571366 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04464849 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04534512 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04406613 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571319 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571360 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04296358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04380175 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571316 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571363 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773087 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571309 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04715759 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571300 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571332 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571266 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04638035 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773082 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571346 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04579539 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571359 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773084 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571342 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04947443 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773096 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571270 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571373 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04743805 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04715758 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571304 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571299 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571308 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04773093 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23571351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04877398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04638032 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10528955 - -
Affymetrix Mouse Gene 1.0 ST Array 10528953 - -
Affymetrix Mouse Gene 1.0 ST Array 10528966 - -
Affymetrix Mouse Gene 1.0 ST Array 10528965 - -
Affymetrix Mouse Gene 1.0 ST Array 10528964 - -
Affymetrix Mouse Gene 1.0 ST Array 10528952 - -
Affymetrix Mouse Gene 1.0 ST Array 10528960 - -
Affymetrix Mouse Gene 1.0 ST Array 10528957 - -
Affymetrix Mouse Gene 1.0 ST Array 10528956 - -
Affymetrix Mouse Gene 1.0 ST Array 10528968 - -
Affymetrix Mouse Gene 1.0 ST Array 10528958 - -
Affymetrix Mouse Gene 1.0 ST Array 10528959 - -
Affymetrix Mouse Gene 1.0 ST Array 10528961 - -
Affymetrix Mouse Gene 1.0 ST Array 10528962 - -
Affymetrix Mouse Gene 1.1 ST Array 10528951 - -
Affymetrix Mouse Clariom S Array PSR0500019551.mm.1 TC0500002155.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019551.mm.1 NM_001014394.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019551.mm.1 NM_001014394.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 767448 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_2529810 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2537217 - -
MouseRef-8 v2 Expression BeadChip ILMN_2909568 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2909568 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0