This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01221765_m1
See other ADGRF3 GE Assays ›
Gene Symbol:
Adgrf3
Gene Name:
adhesion G protein-coupled receptor F3
Gene Aliases:
Gm1041, Gpr113, Pgr23
Chromosome Location:
Chr.5: 30193431 - 30205754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01221765_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014394.2 NP_001014416.2 5 - 6 699 56
GenBank mRNA AY701869.1 - 5 - 6 699 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002155.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrf3

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019563.mm.1 NM_001014394.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019563.mm.1 NM_001014394.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019563.mm.1 TC0500002155.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019562.mm.1 NM_001014394.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019562.mm.1 NM_001014394.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019562.mm.1 TC0500002155.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline