This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01221885_m1
See other CHAT GE Assays ›
Gene Symbol:
Chat
Gene Name:
choline acetyltransferase
Gene Aliases:
B230380D24Rik
Chromosome Location:
Chr.14: 32408203 - 32465909 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01221885_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009891.2 NP_034021.1 6 - 7 765 68
GenBank mRNA AK046406.1 - 6 - 7 766 68
AK047708.1 - 5 - 6 657 68
BC119322.1 - 5 - 6 527 8734071 68
BC120589.1 - 5 - 6 527 40129512 68
D12487.1 - 5 - 6 689 68
D12490.1 - 5 - 6 658 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001703.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chat

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

chemical synaptic transmission
neuromuscular synaptic transmission
muscle organ development
establishment of synaptic specificity at neuromuscular junction
memory
rhythmic behavior
adult walking behavior
acetylcholine biosynthetic process
dendrite development
neuron differentiation
neurotransmitter biosynthetic process
rhythmic excitation
choline O-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
choline binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0