This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222529_m1
See other RGR GE Assays ›
Gene Symbol:
Rgr
Gene Name:
retinal G protein coupled receptor
Gene Aliases:
-
Chromosome Location:
Chr.14: 37034909 - 37049014 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222529_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301692.1 NP_001288621.1 3 - 4 471 114
NM_001301694.1 NP_001288623.1 3 - 4 421 114
NM_021340.4 NP_067315.1 4 - 5 593 114
GenBank mRNA AF076930.1 - 4 - 5 513 114
AK044450.1 - 4 - 5 545 114
AK044649.1 - 4 - 5 544 114
BC082611.1 - 4 - 5 530 114
BC086689.1 - 4 - 5 520 114
BC119055.1 - 4 - 5 527 8733804 114
CK623457.1 - 3 - 4 354 112
CK626179.1 - 3 - 4 409 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgr

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
signal transduction
G-protein coupled receptor signaling pathway
visual perception
phototransduction
protein-chromophore linkage
response to stimulus
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
photoreceptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0