This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222535_m1
See other CDHR1 GE Assays ›
Gene Symbol:
Cdhr1
Gene Name:
cadherin-related family member 1
Gene Aliases:
Pcdh21, Prcad, mKIAA1775
Chromosome Location:
Chr.14: 37077849 - 37098311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222535_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdhr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130878.2 NP_570948.1 12 - 13 1493 94
GenBank mRNA AB093300.1 - 12 - 13 1909 94
AF426393.1 - 12 - 13 1380 94
AK143151.1 - 12 - 13 1434 94
BC042454.1 - 12 - 13 1403 5369576 94
BC058270.1 - 12 - 13 1493 6490597 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001745.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdhr1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular process
photoreceptor cell maintenance
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013582.mm.1 NM_130878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013582.mm.1 NM_130878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400013582.mm.1 TC1400001745.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline