This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222874_g1
See other PPP2R3D GE Assays ›
Gene Symbol:
Ppp2r3d
Gene Name:
protein phosphatase 2 (formerly 2A), regulatory subunit B'', delta
Gene Aliases:
PP2A B''-PR59, PP2A PR59, PR59, Ppp2r3, Ppp2r3a
Chromosome Location:
Chr.9: 124438769 - 124441010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222874_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011154.2 NP_035284.2 364 123
NR_028110.2 364 123
NR_102716.1 364 123
GenBank mRNA AK054122.1 - 364 123
AK143212.1 - 152 123

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK143212.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r3d

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of cell proliferation
regulation of protein phosphatase type 2A activity
negative regulation of G1/S transition of mitotic cell cycle
chromatin binding
protein binding
protein phosphatase type 2A regulator activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NM_001163415.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NM_001163415.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NM_011154.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NM_011154.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NR_028110.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 NR_102716.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900028027.mm.1 TC0900003321.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0