Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400001033.mm.1 |
NM_025476.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001033.mm.1 |
NM_025476.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001033.mm.1 |
TC0400000164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001025.mm.1 |
NM_025476.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001025.mm.1 |
NM_025476.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001025.mm.1 |
TC0400000164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001026.mm.1 |
NM_025476.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001026.mm.1 |
NM_025476.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001026.mm.1 |
TC0400000164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001029.mm.1 |
NM_025476.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001029.mm.1 |
NM_025476.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001029.mm.1 |
TC0400000164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001035.mm.1 |
NM_025476.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001035.mm.1 |
NM_025476.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001035.mm.1 |
TC0400000164.mm.1 |
MTA TC |