This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222983_m1
See other RMDN1 GE Assays ›
Gene Symbol:
Rmdn1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
Fam82b, RMD-1
Chromosome Location:
Chr.4: 19574862 - 19606932 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222983_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rmdn1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025476.6 NP_079752.3 4 - 5 632 121
GenBank mRNA AK002442.1 - 4 - 5 515 121
AK009059.1 - 4 - 5 490 121
AK010421.2 - 4 - 5 509 121
AK029446.1 - 4 - 5 492 121
AK160266.1 - 4 - 5 513 121
BC025158.1 - 4 - 5 505 4023319 121
BC096046.1 - 4 - 5 609 1397855 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000164.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rmdn1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001037.mm.1 NM_025476.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001037.mm.1 NM_025476.6 Refseq
Affymetrix Mouse Clariom S Array PSR0400001037.mm.1 TC0400000164.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-m6z7x:80/100.66.72.99:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0