This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223038_m1
See other TMPRSS11B GE Assays ›
Gene Symbol:
Tmprss11b
Gene Name:
transmembrane protease, serine 11B
Gene Aliases:
9930019B18Rik, Tmprss11bnl
Chromosome Location:
Chr.5: 86659522 - 86676298 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223038_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK036858.1 - 80 73
AK036968.1 - 1 - 2 80 73
AK137654.1 - 92 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK036858.1, AK036968.1, AK137654.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 614493 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0