This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223294_m1
See other TMPRSS11C GE Assays ›
Gene Symbol:
Tmprss11c
Gene Name:
transmembrane protease, serine 11c
Gene Aliases:
Tmprss13
Chromosome Location:
Chr.5: 86231481 - 86289308 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223294_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030297.2 NP_001025468.1 4 - 5 413 61
GenBank mRNA AM260217.1 - 4 - 5 413 61
BC147157.1 - 4 - 5 394 9056205 61
BC147158.1 - 4 - 5 394 9056165 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002678.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss11c

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500023638.mm.1 NM_001030297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023638.mm.1 NM_001030297.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500023638.mm.1 TC0500002678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023639.mm.1 NM_001030297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023639.mm.1 NM_001030297.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500023639.mm.1 TC0500002678.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline