This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223534_m1
See other CNTFR GE Assays ›
Gene Symbol:
Cntfr
Gene Name:
ciliary neurotrophic factor receptor
Gene Aliases:
Cntfralpha
Chromosome Location:
Chr.4: 41657498 - 41697091 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223534_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntfr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136056.2 NP_001129528.1 4 - 5 537 102
NM_016673.2 NP_057882.2 4 - 5 516 102
GenBank mRNA AB154246.1 - 2 - 3 312 102
AF068615.1 - 4 - 5 467 102
BC046974.1 - 4 - 5 506 6389656 102
BC050928.1 - 4 - 5 534 6415187 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002475.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntfr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
brainstem development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
ciliary neurotrophic factor-mediated signaling pathway
cytokine receptor activity
ciliary neurotrophic factor receptor activity
receptor binding
protein binding
cytokine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400019987.mm.1 NM_001136056.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400019987.mm.1 NM_016673.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019987.mm.1 NM_016673.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400019987.mm.1 TC0400002475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0