This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223611_m1
See other PCMTD1 GE Assays ›
Gene Symbol:
Pcmtd1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
8430411F12Rik, A030012M09Rik
Chromosome Location:
Chr.1: 7088891 - 7173628 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223611_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcmtd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183028.3 NP_898849.1 1 - 2 294 97
GenBank mRNA AK166166.1 - 1 - 2 179 97
AK168606.1 - 1 - 2 46 97
AK168705.1 - 1 - 2 64 97
BC019193.1 - 1 - 2 43 97
BC110360.1 - 1 - 2 64 5326388 97
BY282003.1 - 1 - 2 295 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK166166.1, AK168606.1, AK168705.1, BC110360.1, NM_183028.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcmtd1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
biological_process
molecular_function
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100000247.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000247.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000247.mm.1 TC0100000027.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100000253.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000253.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000253.mm.1 TC0100000027.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100000248.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000248.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000248.mm.1 TC0100000027.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100000254.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000254.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000254.mm.1 TC0100000027.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100000251.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000251.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000251.mm.1 TC0100000027.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 496940 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-66ncq:80/100.66.77.55:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0