This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223613_g1
See other PCMTD1 GE Assays ›
Gene Symbol:
Pcmtd1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
8430411F12Rik, A030012M09Rik
Chromosome Location:
Chr.1: 7088891 - 7173628 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223613_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcmtd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183028.3 NP_898849.1 3 - 4 812 89
GenBank mRNA AK040848.1 - 3 - 4 540 89
AK079468.1 - 2 - 3 349 89
AK145008.1 - 3 - 4 538 89
AK166166.1 - 3 - 4 697 89
AK166593.1 - 3 - 4 538 89
AK168606.1 - 3 - 4 564 89
AK168705.1 - 3 - 4 582 89
BC019193.1 - 3 - 4 560 89
BC052741.1 - 2 - 3 529 6833458 89
BC110360.1 - 3 - 4 582 5326388 89
BC117941.1 - 2 - 3 529 40097836 89
BC117942.1 - 2 - 3 529 40097839 89

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcmtd1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
biological_process
molecular_function
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100000261.mm.1 NM_183028.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000261.mm.1 NM_183028.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100000261.mm.1 TC0100000027.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0