This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223801_g1
See other STARD8 GE Assays ›
Gene Symbol:
Stard8
Gene Name:
START domain containing 8
Gene Aliases:
Dlc3, mKIAA0189
Chromosome Location:
Chr.X: 99003228 - 99074728 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223801_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199018.2 NP_950183.1 12 - 13 3139 89
GenBank mRNA AK133927.1 - 12 - 13 3087 89
AK149830.1 - 12 - 13 3070 89
AK150259.1 - 12 - 13 3070 89
AK172910.1 - 3 - 4 561 89
BC013223.1 - 2 - 3 916 89
BC023434.1 - 4 - 5 663 89
BC034186.1 - 8 - 9 2261 89
BC062944.1 - 12 - 13 3054 30357177 89
BC078461.1 - 11 - 12 2902 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000933.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard8

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006044.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006044.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006044.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006045.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006045.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006045.mm.1 TC0X00000933.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0