This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223804_m1
See other STARD8 GE Assays ›
Gene Symbol:
Stard8
Gene Name:
START domain containing 8
Gene Aliases:
Dlc3, mKIAA0189
Chromosome Location:
Chr.X: 99003228 - 99074728 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223804_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199018.2 NP_950183.1 3 - 4 367 131
GenBank mRNA AK133927.1 - 3 - 4 315 131
AK149830.1 - 3 - 4 298 131
BC062944.1 - 3 - 4 282 30357177 131
BC078461.1 - 2 - 3 132 131

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK133927.1, AK149830.1, BC062944.1, NM_199018.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard8

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity
lipid binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006023.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006023.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006023.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006024.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006024.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006024.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006030.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006030.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006030.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006029.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006029.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006029.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006025.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006025.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006025.mm.1 TC0X00000933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006031.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006031.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006031.mm.1 TC0X00000933.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 601354 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0