This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223808_g1
See other STARD8 GE Assays ›
Gene Symbol:
Stard8
Gene Name:
START domain containing 8
Gene Aliases:
Dlc3, mKIAA0189
Chromosome Location:
Chr.X: 99003228 - 99074728 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223808_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199018.2 NP_950183.1 7 - 8 2170 68
GenBank mRNA AK133927.1 - 7 - 8 2118 68
AK149830.1 - 7 - 8 2101 68
AK150259.1 - 7 - 8 2101 68
BC034186.1 - 3 - 4 1292 68
BC062944.1 - 7 - 8 2085 30357177 68
BC078461.1 - 6 - 7 1933 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000933.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard8

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006038.mm.1 NM_199018.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006038.mm.1 NM_199018.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006038.mm.1 TC0X00000933.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0