This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224332_m1
See other BORA GE Assays ›
Gene Symbol:
Bora
Gene Name:
bora, aurora kinase A activator
Gene Aliases:
6720463M24Rik, AI317232
Chromosome Location:
Chr.14: 99046247 - 99076450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224332_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bora
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175265.4 NP_780474.1 6 - 7 771 93
GenBank mRNA AK034930.1 - 6 - 7 772 93
AK141389.1 - 6 - 7 722 93
AK167430.1 - 6 - 7 703 93
BC052183.1 - 6 - 7 672 6336038 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001152.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bora

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of mitotic nuclear division
regulation of protein localization
cell division
regulation of mitotic spindle organization
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009061.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009061.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009061.mm.1 TC1400001152.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0