This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224814_m1
See other SLCO2A1 GE Assays ›
Gene Symbol:
Slco2a1
Gene Name:
solute carrier organic anion transporter family, member 2a1
Gene Aliases:
2310021C19Rik, Pgt, Slc21a2, mPgt
Chromosome Location:
Chr.9: 103007668 - 103087849 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224814_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco2a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033314.3 NP_201571.2 10 - 11 1580 71
GenBank mRNA AF323958.1 - 10 - 11 1449 71
AK087350.1 - 10 - 11 1587 71
AK166724.1 - 10 - 11 1581 71
AK169706.1 - 10 - 11 1583 71
BC035200.1 - 1380 3158233 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001297.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco2a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
prostaglandin transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
prostaglandin transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010094.mm.1 NM_033314.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010094.mm.1 NM_033314.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900010094.mm.1 TC0900001297.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0