This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224902_m1
See other GABRA1 GE Assays ›
Gene Symbol:
Gabra1
Gene Name:
gamma-aminobutyric acid (GABA) A receptor, subunit alpha 1
Gene Aliases:
Gabra-1
Chromosome Location:
Chr.11: 42130939 - 42182951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224902_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabra1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010250.5 NP_034380.1 6 - 7 1334 65
GenBank mRNA AK034095.1 - 6 - 7 1335 65
AK039496.1 - 6 - 7 618 65
AK141596.1 - 6 - 7 1194 65
BC132331.1 - 6 - 7 1010 40130608 65
BC137733.1 - 6 - 7 1013 8860752 65
M63436.1 - 6 - 7 1007 65
M86566.1 - 6 - 7 702 65
X61430.1 - 5 - 6 590 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002557.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabra1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
gamma-aminobutyric acid signaling pathway
synaptic transmission, GABAergic
cellular response to histamine
GABA-A receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
drug binding
GABA receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline