This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224982_m1
See other HDAC8 GE Assays ›
Gene Symbol:
Hdac8
Gene Name:
histone deacetylase 8
Gene Aliases:
2610007D20Rik
Chromosome Location:
Chr.X: 102284640 - 102509686 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224982_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027382.3 NP_081658.1 4 - 5 515 97
GenBank mRNA AK011332.1 - 4 - 5 516 97
AK034511.1 - 4 - 5 481 97
AK041965.1 - 4 - 5 514 97
AK131998.1 - 4 - 5 728 97
BC061257.1 - 4 - 5 487 30251620 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002741.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac8

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
sister chromatid cohesion
chromatin modification
histone deacetylation
regulation of cohesin loading
histone deacetylase activity
transcription factor binding
hydrolase activity
histone deacetylase activity (H3-K14 specific)
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
histone deacetylase activity (H4-K16 specific)
metal ion binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
NAD-dependent histone deacetylase activity (H3-K18 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00017302.mm.1 NM_027382.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017302.mm.1 TC0X00002741.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00017301.mm.1 NM_027382.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017301.mm.1 TC0X00002741.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0