This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225045_m1
See other AWAT1 GE Assays ›
Gene Symbol:
Awat1
Gene Name:
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases:
Dgat2l3
Chromosome Location:
Chr.X: 100572247 - 100578206 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225045_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Awat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081136.1 NP_001074605.1 4 - 5 546 74
GenBank mRNA AK137763.1 - 4 - 5 543 74
BC147680.1 - 4 - 5 517 9056653 74
BC147683.1 - 4 - 5 517 9056656 74
BC147829.1 - 4 - 5 475 9087546 74
BC147832.1 - 4 - 5 475 9087549 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000954.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
molecular_function
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
long-chain-alcohol O-fatty-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006155.mm.1 NM_001081136.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006155.mm.1 NM_001081136.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006155.mm.1 TC0X00000954.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0