This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225232_m1
See other TFDP2 GE Assays ›
Gene Symbol:
Tfdp2
Gene Name:
transcription factor Dp 2
Gene Aliases:
1110029I05Rik, A330080J22Rik, DP-3, DP3
Chromosome Location:
Chr.9: 96196275 - 96323646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225232_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfdp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001184706.1 NP_001171635.1 2 - 3 339 140
NM_001184708.1 NP_001171637.1 2 - 3 339 140
GenBank mRNA AK029055.1 - 2 - 3 312 140
AK044784.1 - 2 - 3 341 140
AK053881.1 - 2 - 3 297 140

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK029055.1, AK044784.1, NM_001184706.1, NM_001184708.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Apoptosis Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell cycle
negative regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
protein binding
transcription factor binding
protein domain specific binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900009536.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009536.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009536.mm.1 NM_001184708.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009536.mm.1 NM_001184708.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009536.mm.1 TC0900001210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184709.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_001184711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_178667.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 NM_178667.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900009543.mm.1 TC0900001210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_001184711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_001184711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_178667.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 NM_178667.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900009542.mm.1 TC0900001210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184708.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184708.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184709.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184710.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184710.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_001184711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_178667.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 NM_178667.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900009548.mm.1 TC0900001210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184709.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184710.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184710.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_001184711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_178667.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 NM_178667.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900009544.mm.1 TC0900001210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184709.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184710.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184710.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_001184711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_178667.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 NM_178667.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900009546.mm.1 TC0900001210.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0