This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225578_m1
See other UNC50 GE Assays ›
Gene Symbol:
Unc50
Gene Name:
unc-50 homolog (C. elegans)
Gene Aliases:
1110002A21Rik, GMH1, UNCL, URP
Chromosome Location:
Chr.1: 37430144 - 37439124 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225578_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026123.3 NP_080399.1 3 - 4 729 65
GenBank mRNA AK003273.1 - 3 - 4 504 65
AK004433.1 - 3 - 4 493 65
AK132757.1 - 3 - 4 730 65
AK135885.1 - 3 - 4 721 65
AY017214.1 - 3 - 4 714 65
BC019484.1 - 3 - 4 696 4039903 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000260.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc50

Gene Ontology Categories:

Function(s) Process(es)

transport
cell surface receptor signaling pathway
protein transport
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100001858.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001858.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001858.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001852.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001852.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001852.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001854.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001854.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001854.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001857.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001857.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001857.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001855.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001855.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001855.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001853.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001853.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001853.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001856.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001856.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001856.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001859.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001859.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001859.mm.1 TC0100000260.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0