Assay Details
| Gene Symbol: Unc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026123.3 | NP_080399.1 | 4 - 5 | 867 | 110 | |
| GenBank mRNA | AK003273.1 | - | 4 - 5 | 642 | 110 | |
| AK004433.1 | - | 4 - 5 | 631 | 110 | ||
| AK132757.1 | - | 4 - 5 | 868 | 110 | ||
| AK135885.1 | - | 4 - 5 | 859 | 110 | ||
| AK210936.1 | - | 1 - 2 | 45 | 110 | ||
| AY017214.1 | - | 4 - 5 | 852 | 110 | ||
| BC019484.1 | - | 4 - 5 | 834 | 4039903 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc50
Genomic Map