This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225787_m1
See other NDUFAF7 GE Assays ›
Gene Symbol:
Ndufaf7
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7
Gene Aliases:
2410091C18Rik, AL033374, PRO1853
Chromosome Location:
Chr.17: 78937106 - 78948052 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225787_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufaf7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028611.3 NP_082887.2 7 - 8 798 71
GenBank mRNA AK010752.1 - 7 - 8 797 71
AK090097.1 - 7 - 8 797 71
BC108350.1 - 7 - 8 785 4220568 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001192.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufaf7

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial respiratory chain complex I assembly
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011271.mm.1 NM_028611.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011271.mm.1 NM_028611.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700011271.mm.1 TC1700001192.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0